Masterflex® L/S® Digital Pump Drives
8/25/2008
New graphical LCD interface offers more intuitive setup and operation
With their well-deserved reputation for durability and ease of use, NEW MASTERFLEX L/S digital drives have moved one step further with an evolutionary redesign. These precision digital drives now offer high-accuracy motors and an all-new graphical interface that makes setup and operation easier than ever. The high-contrast graphical LCD shows four operating modes: continuous run, timed dispense, copy dispense, and volume dispense allowing you to view pump performance continually. The maintenance-free brushless motor offers ±0.1% speed control accuracy with a 6000:1 turndown ratio for a wider flow range. The menu-driven interface lets you select one of seven languages for programming and operation.
Enhanced dispensing features include volume dispense, timed dispense, and copy/repeat. A programmable dispense interval allows hands-free dispensing and filling. Cumulative volume feature totalizes volume dispensed over multiple cycles. Remote control capabilities allow easy integration into automated processes. Available models include bi-directional RS-232 or USB connectivity for control through a PC, and an IP66-rated all-stainless steel housing for use in sanitary, washdown applications. New MASTERFLEX L/S digital pump drives are compatible with all existing L/S pump heads.