Ideal for pharmaceutical, food & beverage, chemical, and water treatment applications
Cole-Parmer Instrument Company features two of the most precise and reliable digital, peristaltic tubing pump drives on the market-the L/S 07575-00 and the I/P 77420-00.
These stainless steel drives are ideally suited for accurate fluid dispensing, metering, and transfer applications in harsh or demanding environments. Both drives are IP66 (NEMA 4X) rated, and feature fully sealed 316 stainless steel housings that are easy to clean and resistant to commonly used cleaning and sanitizing solutions. The reversible, maintenance-free brushless motors offer full remote control capabilities, allowing easy integration into your automated processes.
Flow range for the L/Sr drive is 0.1 to 3400 mL/min with a speed turndown of 600:1; flow range for the I/Pr drive is 0.01 to 26 LPM with a turndown of 650:1 (flow rates depend on motor rpm and tubing size). Speed control accuracy is ±0.25%.
The large LED display on both models gives you a clear, digital readout of motor rpm, flow rate, dispense volume, and dispense copy number. The sealed membrane keypad allows for easy programming of all parameters, while the keypad's lock/unlock feature protects settings.
Drives accept all Masterflex L/Sr and I/Pr pump heads, and all L/Sr and I/Pr pump tubing formulations, respectively.
Download 300 DPI 07575APP13RIGHT.jpg Image