Masterflex® Chem-Durance™ Tubing
6/2/2006
Protective inner liner-flexible outer layer
Cole-Parmer presents Chem-Durance, an exclusive, all-new tubing formulation for the MASTERFLEX® peristaltic pump series. This co-extruded material has an inner liner that protects against chemically aggressive fluids, and a soft, flexible outer layer that allows it to be used in a peristaltic pump.
Not only does Precision Chem-Durance tubing have great chemical compatibility, it also has excellent tubing life and durability, especially when operating under pressure. There is minimal spallation of the outer coating, requiring less maintenance on the pump. The flexible outer layer also allows for easier handling and loading into the pump head.
Precision Chem-Durance 06432-series tubing is available in the MASTERFLEXr L/Sr and I/Pr sizes. Like other MASTERFLEXr tubing formulations, Chem-Durance is manufactured to extremely close tolerances ensuring accurate and repeatable flow.