Flow tube and hood molded of impact-resistant, medical-grade Lexan®
Available for use with oxygen and breathing air, these flowmeters feature a precision control valve for accurate, leak-free flow control and are manufactured with a machined brass base with integral DISS fitting and plated with a high-gloss chrome-like finish.
These low-cost flowmeters are perfect for clinical devices that regulate oxygen and air into processes. All flowmeters are made with a high impact-resistant Lexan® shield and flow tube for safety and durability. Combined with a sturdy machined brass base, these meters will stand up to the rigors of clinical or field use. The durable, stainless steel stem valve ensures precise control throughout the full flow range and provides for positive flow shutoff. O-ring seals eliminate the leaks that are commonly associated with plastic-base models. Meters are calibrated at 50 psig inlet pressure for accurate flow measurement under a variety of operating conditions.
Download 300 DPI 32900_62.jpg Image