Innovative cameras provide views into cramped and narrow spaces for inspection and analysis
Vernon Hills, Ill., August 26, 2010—An economical new video borescope inspection camera takes users where they haven’t been before. The miniature waterproof camera head allows high-resolution viewing in small spaces—ideal for automotive applications, building maintenance, and laboratory analysis.
The Extech Video Borescope Inspection Camera features a 17-mm diameter camera head with four built-in illuminating LEDs and a 2.4" color TFT LCD monitor. Video can be output for viewing on any monitor with a video input jack. Images are glare-free even with close-up viewing. A 39"L flexible gooseneck cable retains its configured shape.
A wireless version is also available with a 9-mm diameter camera head and a detachable 3.5" color TFT LCD wireless monitor with 2GB microSD memory card for capturing images (JPEG) or video (AVI) for viewing on your PC. The wireless LCD can be viewed from a remote location up to 32 feet from measurement point. Its menu displays 10 languages.
To find out more about the Extech Video Borescope Inspection Cameras from Cole-Parmer, call 800-323-4340 or visit ColeParmer.com/8060.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the U.S., call 800-323-4340. International customers, call 847-549-7600. Visit us at www.coleparmer.com.
Download 300 DPI 68517_10.jpg Image