Eutech 2000D Differential pH Controller
8/25/2004
Greater accuracy and reliability
Eutechr Instruments' new 1/2-DIN pH 2000D controller takes advantage of differential probe technology to provide added reliability in pharmaceutical and industrial applications.
Differential pH measurement reduces reference junction fouling and virtually eliminates ground loops in the most demanding applications. Three front-panel selectable control modes (limit, proportional, and proportional-integral) with separately adjustable set points, delay, and hysteresis provide precise high- and low-level control. Two control relays along with separate alarm and wash relays offer flexibility to handle almost any application. The back-lit LCD indicates pH and temperature values that can be recorded via two, galvanically isolated 0/4 to 20 mA outputs.
Download 300 DPI 5671520.jpg Image