I/P Digital Peristaltic Pump Drive Designed for manufacturing processes, quick fluid transfer in the lab, and industrial applications
Designed for manufacturing processes, quick fluid transfer in the lab, and industrial applications, Masterflex® I/P® Digital Modular Dispensing Drives from Cole-Parmer are precision peristaltic pump drives with flow rates ranging from 0.0006 LPM up to 26 LPM.
The modular design allows the controller be placed separately from the pump motor for optimal convenience and operator safety. Three dispensing modes and low- maintenance motors add to their flexibility. Automated dispensing by time and volume is easy to achieve with user- friendly interface. A programmable anti-drip function briefly reverses the motor at end of the cycle to increase dispense accuracy and prevent dripping.
Two models available; a benchtop controller unit or a washdown, wall-mount unit. Call 020- 85747556 or visit ColeParmer.co.uk/21419.
Download 300 DPI 07594_10.jpg Image