Economical Conductivity Monitor and Monitor/Controller
11/10/2005
Local monitoring or control without the expense of mA outputs
The new, IP65-rated CON 550 monitor and CON 560 monitor/controller from Eutech Instruments, display conductivity and temperature values, and feature manual or automatic temperature compensation for accurate industrial measurement. Units accept two- or four-cell conductivity electrodes.
The CON 550 is an inexpensive local monitor with display. The CON 560 controller/monitor features a backlit display and two relays with separately adjustable set points, hysteresis, and delay for on/off control. Both models are powered by 110 VAC to 9 VDC converters.