These economical electronic paperless recorders display real-time data in digital or trend formats, and offer two universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTDs, frequency, and pulse inputs. Recorders are available with USB 2.0 or 10/100Base T Ethernet communication protocols, providing additional flexibility for monitoring conditions in all of your process environments.
Units allow you to store up to 2 gigabytes of data on optional CompactFlash™ cards, or easily manage your data by downloading to an Excel® file with the included Navigatorlite™ software. Utilize the optional Navigator™ software to review, print, and export your data displaying your scales, including date-and-time stamp information, for all of your data monitoring requirements.