Cole-Parmer Roto-Torque® Rotator
5/10/2007
Gentle but efficient mixing
Cole-Parmer proudly offers a proprietary rotator designed to efficiently mix the contents of test tubes, flasks, and bottles for general laboratory applications.
Unit features a variable speed from 3 to 80 rpm, providing gentle to vigorous agitation. Adjustable rotation angle controls the amount of mixing; rotate samples horizontally for minimal rolling, vertically for end-over-end mixing, or anywhere in between. Quick and easy loading and removal of sample vessels reduce the chance of accidental spills and breakage.
Rotator operates in temperatures up to 40°C and relative humidity ranging from 20 to 80%- making this rotator great for use in incubators. Also ideal for applications involving aerating cultures, keeping biological samples in suspension, liquid extractions, hybridizations, preparation of homogenous dispersions, and general mixing applications.