Wider selection of power supplies to fit all your applications
Cole-Parmer Instrument Company announces the addition of B&K Precision power supplies to its recently expanded DC Power Supply product line. New offerings include single-output, triple-output, and high-current power supplies.
Single-output units from B&K Precision feature an adjustable output with overload protection, constant current, and constant voltage operation. Select from benchtop models (26868-00 through -18) or rack-mount models (26868-40 and -42).
Triple-output power supplies provide multiple outputs for greater application versatility. Model 26868-30 features two fixed and one adjustable output, model 26868-32 features one fixed and two adjustable outputs, and models 26868-34 and -36 feature three adjustable outputs.| High-current power supplies (26868-20 and -22) operate continuously at full load without overheating. The rugged design withstands current limiting overload, overvoltage, short circuits, and reverse polarity.
Download 300 DPI 2686830.jpg Image