Precision-adjusting valve for accurate, leak-free flow control
Cole-Parmer introduces a new line of impact-resistant polycarbonate flowmeters designed for noncaustic liquids and air. Direct-reading flowmeter models 32900-02 to -54 are economically priced and feature scales in both English and metric units. They have a 10:1 turndown ratio, and are designed to maintain pressures to 100 psig and temperatures to 150°F. Liquid flow range is 0.2 to 40 GPH. Air flow range is 0.2 to 60 scfh. Flowmeter accuracy is ±4% full-scale.