Modular mixer features dual functioning keypads
Cole-Parmer introduces a unique, industry-changing overhead mixer with many exciting new features. With such innovations as a full-function remote control keypad and lockout feature, this mixer system is sure to change the way you look at mixing.
This modular unit, exclusive to Cole-Parmer, offers the option of dual keypads, one on the mixer head itself, and a remote that can be used up to six feet away. Both keypads control all mixing functions simultaneously. Use the remote to operate a mixer from even outside of a fume hood—attach controller to the hood with the use of onboard magnets.
A programmable reversing feature alters mixing direction, homogenizing materials more quickly. A timed cycle feature turns the mixer off once a programmed cycle is complete. Use both features simultaneously to reduce downtime and the need to monitor cycles.
Eliminate inadvertent changes in speed and cycle times with an easy-to-use lockout function. The integrated sample light illuminates your work even in the worst lighting conditions. All this packed into a compact and lightweight design.