This product is now discontinued, View Related Products

Aquamax Titrator Accessories

Keep your titrator working at its best with these replacement parts and accessories.
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Aquamax KF Plus Titrator Syringe Assembly; 12.5 mL WZ-94105-01 Mfr # 594013
COMPARE
Titrator Syringe Assembly; 12.5 mL
Call For Price
Aquamax KF Plus Titrator Dispensing Tip WZ-94105-02 Mfr # 594014
COMPARE
Titrator Dispensing Tip
Call For Price
Aquamax KF Plus Titrator Electrode; Non-Aqueous with Cable WZ-94105-03 Mfr # 594033
COMPARE
Titrator Electrode; Non-Aqueous with Cable
Call For Price
Aquamax KF Plus Titrator Electrode; Aqueous with Cable WZ-94105-04 Mfr # 594034
COMPARE
Titrator Electrode; Aqueous with Cable
Call For Price
Aquamax KF Plus Titrator Electrode; Precipitation with Cable WZ-94105-05 Mfr # 594063
COMPARE
Titrator Electrode; Precipitation with Cable
Call For Price
Aquamax KF Plus Titrator Electrode; Chloride with Cable WZ-94105-06 Mfr # 594064
COMPARE
Titrator Electrode; Chloride with Cable
Call For Price
Aquamax KF Plus Titrator Titration Vessel; 170 mL WZ-94105-07 Mfr # 301802
COMPARE
Titrator Titration Vessel; 170 mL
Call For Price
Aquamax KF Plus Titrator Titration Vessel; 210 mL WZ-94105-08 Mfr # 594043
COMPARE
Titrator Titration Vessel; 210 mL
Call For Price
Aquamax KF Plus Titrator Electrode Platform with Cables WZ-94105-09 Mfr # 594006
COMPARE
Titrator Electrode Platform with Cables
Call For Price
Aquamax KF Plus Titrator Syringe Drive Burette with Cables WZ-94105-10 Mfr # 594007
COMPARE
Titrator Syringe Drive Burette with Cables
Call For Price
Aquamax KF Plus Titrator Drying Tube with Desiccant WZ-94105-11 Mfr # 594020
COMPARE
Titrator Drying Tube with Desiccant
Call For Price
Aquamax KF Plus Titrator 12.5 mL Syringe WZ-94105-12 Mfr # 594017
COMPARE
Titrator 12.5 mL Syringe
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat