This product is now discontinued, View Related Products
YSI Model 5000 Dissolved Oxygen Benchtop Meters
Keypad with tactile keys provide audible confirmations to reduce errors
  • Large LCD is easy to read—even from 10 feet away
  • Menu-driven operation makes setup simple
These benchtop meters are easy to use. Meters feature autocalibration, data storage of up to 100 sets, automatic stabilization, and real-time clock. Meters operate on AC power or batteries. Order probes and accessories separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Dissolved Oxygen Min Dissolved Oxygen
Availability Pricing
YSI 5000-115 Model 5000 Benchtop Dissolved Oxygen Meter, 115 VAC WZ-05510-20 Mfr # 050022
COMPARE
0.0 to 60.0 ppm
Call For Price
YSI 050023 Benchtop Dissolved Oxygen Meter, 230 V WZ-05510-25 Mfr # 050023
COMPARE
0.0 to 60.0 ppm