YSI DO Cap Membrane Kit
Makes membrane chanages quick and easy
  • Teflon caps offer reliable performance for most dissolved oxygen applications
  • Provides superb fouling resistance
The DO cap membrane kit is ideal for use with the YSI DO probes and DO meters. It features a fouling resistance with a response time of 18 seconds and flow dependence of 60%.
£75.60GBP / Each
£90.72GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
YSI 5906 DO Cap Membrane Kit; 6 Black Screw-on Cap Membranes (1 mil PTFE), Bottle of Electrolyte, and Sanding Disc WZ-05519-61 Mfr # 059880
COMPARE
YSI 5239, 5905, 5010 DO Probes and the 556 and 85 DO meters Replacement DO membrane cap kit, 1.00 mil PTFE
£75.60GBP / Each
£90.72GBP (incl VAT)