YSI Self-Stirring BOD Probes

Ensures proper sample agitation
  • Fits 300 mL BOD bottles with 5-degree taper
  • EPA approved methodology
These BOD probes provide continuous stirring for accurate measurement and faster response time. They connect directly to YSI 5000/5100 benchtop dissolved oxygen meters (adapter need for model 05519-42). They are EPA approved for wastewater compliance reporting of BOD and CBOD.
£756.27 - £810.93GBP / Each
£907.52 - £973.12GBP (incl VAT)

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI 5010 Self-stirring BOD probe, direct connect to 5000/5100 meters WZ-05510-60 Mfr # 050102
COMPARE
Self-stirring BOD probe, direct connect to 5000/5100 meters
£756.27GBP / Each
£907.52GBP (incl VAT)
YSI 5905-115 Self-stirring BOD bottle probe for meters;  115 VAC WZ-05519-42 Mfr # 059800
COMPARE
Self-stirring BOD bottle probe, 115 VAC, for meters
£810.93GBP / Each
£973.12GBP (incl VAT)
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat