This product is now discontinued, View Related Products
WTW InoLab Conductivity 7110 and Conductivity 7310 Benchtop Meters
Advanced technology delivers more convenient and reliable measurements
  • AutoRead function ensures repeatable results
  • Easy to calibrate and maintain with calibration timer
  • Power with universal AC adapter or AA batteries
The inoLab family of benchtop meters offer an intuitive user interface, large informative display, and keypad with tactile feedback to provide an exceptional user experience for error-free operation.
inoLab Conductivity 7110 Basic Meter is ideally suited for routine, general laboratory conductivity, TDS, salinity and resistivity measurements. An intuitive user interface and easy-to-clean housing make it ideal for any environment.
inoLab Conductivity 7310 Advanced Meter allows accurate measurements with compliant documentation to support GLP (Good Laboratory Practice) and AQA (Analytical Quality Assurance) laboratory requirements. Capture 500 manual or 5000 automatic data sets with time/date stamp, sample ID, and calibration data. USB interface provides an easy connection to a PC, or choose the model with integrated printer for immediate results. All data is transfered in a CSV format directly into Excel®.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Conductivity Resolution Min Salinity Description
Availability Pricing
WTW COND 7110 inoLab 7110 basic conductivity benchtop meter WZ-19706-02 Mfr # 1CA100
COMPARE
0.001, 0.01, 0.1, or 1 µS; 0.01, 0.1, or 1 mS 0 to 70 ppt inoLab 7110 basic conductivity benchtop meter
Call For Price
WTW COND 7310 inoLab 7310 advanced conductivity benchtop meter WZ-19706-04 Mfr # 1CA300
COMPARE
0.001, 0.01, 0.1, or 1 µS; 0.01, 0.1, or 1 mS 0 to 70 ppt inoLab 7310 advanced conductivity benchtop meter
Call For Price
WTW 1CA300P inoLab 7310 advanced pconductivity benchtop meter with printer WZ-19706-06 Mfr # 1CA300P
COMPARE
0.001, 0.01, 0.1, or 1 µS; 0.01, 0.1, or 1 mS 0 to 70 ppt inoLab 7310 advanced pconductivity benchtop meter with printer