This product is now discontinued, View Related Products
WorkBeads™ Immobilized Metal Ion Affinity Purification Media
High throughput agarose media for capture of His-tagged proteins
  • Cross-linking method results in a highly porous and physically stable matrix
  • Simple coupling procedure provides fast results
  • High chemical stability for easy clean-in-place (CIP)
Separate protein groups on agarose beads with tris(2-aminoethyl)amine (TREN), iminodiacetic acid (IDA), or nitrilotriacetic acid (NTA). Select from uncharged TREN, or charged or uncharged IDA and NTA. Charged media contains IDA or NDA with Ni, Co, Cu, or Zn ions, which are immobilized on the media via the chelating ligand. Metal ion capacity is 50 to 60 µmol/mL.
Call For Price
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
Compareitem specifications
Wetted Materials Media Volume (mL)
Availability Pricing
Bio-Works WorkBeads 40 IDA High Capacity IMAC Medium, Iminodiacetic Acid ; 25 mL WZ-11947-08 Mfr # KN40601001
COMPARE
Iminodiacetic acid (IDA) resin 25
Call For Price
Bio-Works WorkBeads 40 IDA High Capacity IMAC Medium, Iminodiacetic Acid; 150 mL WZ-11947-09 Mfr # KN40601003
COMPARE
Iminodiacetic acid (IDA) resin 150
Call For Price
Bio-Works WorkBeads 40 IDA High Capacity IMAC Medium, Iminodiacetic Acid; 1 L WZ-11947-10 Mfr # KN40601010
COMPARE
Iminodiacetic acid (IDA) resin 1000
Call For Price
Bio-Works WorkBeads 40 NTA IMAC Medium, Nitrilotriacetic Acid; 25 mL WZ-11947-11 Mfr # KN40602001
COMPARE
Nitrilotriacetic acid (NTA) resin 25
Call For Price
Bio-Works WorkBeads 40 NTA IMAC Medium, Nitrilotriacetic Acid; 150 mL WZ-11947-12 Mfr # KN40602003
COMPARE
Nitrilotriacetic acid (NTA) resin 150
Call For Price
Bio-Works WorkBeads 40 NTA IMAC Medium, Nitrilotriacetic Acid; 1 L WZ-11947-13 Mfr # KN40602010
COMPARE
Nitrilotriacetic acid (NTA) resin 1000
Call For Price
Bio-Works WorkBeads 40 TREN High Capacity IMAC Medium, Tris(2-aminoethyl)amine; 25 mL WZ-11947-14 Mfr # KN40603001
COMPARE
Tris(2-aminoethyl)amine (TREN) resin 25
Call For Price
Bio-Works WorkBeads 40 TREN High Capacity IMAC Medium, Tris(2-aminoethyl)amine; 150 mL WZ-11947-15 Mfr # KN40603003
COMPARE
Tris(2-aminoethyl)amine (TREN) resin 150
Call For Price
Bio-Works WorkBeads 40 TREN High Capacity IMAC Medium, Tris(2-aminoethyl)amine; 1 L WZ-11947-16 Mfr # KN40603010
COMPARE
Tris(2-aminoethyl)amine (TREN) resin 1000
Call For Price
Bio-Works WorkBeads 40 Ni High Capacity IMAC Medium for His-Tagged Proteins, Ni-IDA; 25 mL WZ-11947-22 Mfr # KN40650001
COMPARE
Nickel & Iminodiacetic acid resin 25
Call For Price
Bio-Works WorkBeads 40 Ni High Capacity IMAC Medium for His-Tagged Proteins, Ni-IDA; 150 mL WZ-11947-23 Mfr # KN40650003
COMPARE
Nickel & Iminodiacetic acid resin 150
Call For Price
Bio-Works WorkBeads 40 Ni High Capacity IMAC Medium for His-Tagged Proteins, Ni-IDA; 1 L WZ-11947-24 Mfr # KN40650010
COMPARE
Nickel & Iminodiacetic acid resin 1000
Call For Price
Bio-Works WorkBeads 40 Cu High Capacity IMAC Medium for His-Tagged Proteins, Cu-IDA; 25 mL WZ-11947-25 Mfr # KN40650301
COMPARE
Copper & Iminodiacetic acid resin 25
Call For Price
Bio-Works WorkBeads 40 Cu High Capacity IMAC Medium for His-Tagged Proteins, Cu-IDA; 150 mL WZ-11947-26 Mfr # KN40650303
COMPARE
Copper & Iminodiacetic acid resin 150
Call For Price
Bio-Works WorkBeads 40 Cu High Capacity IMAC Medium for His-Tagged Proteins, Cu-IDA; 1 L WZ-11947-27 Mfr # KN40650310
COMPARE
Copper & Iminodiacetic acid resin 1000
Call For Price
Bio-Works WorkBeads 40 Co High Capacity IMAC Medium for His-Tagged Proteins, Co-IDA; 25 mL WZ-11947-28 Mfr # KN40650401
COMPARE
Cobalt & Iminodiacetic acid resin 25
Call For Price
Bio-Works WorkBeads 40 Co High Capacity IMAC Medium for His-Tagged Proteins, Co-IDA; 150 mL WZ-11947-29 Mfr # KN40650403
COMPARE
Cobalt & Iminodiacetic acid resin 150
Call For Price
Bio-Works WorkBeads 40 Co High Capacity IMAC Medium for His-Tagged Proteins, Co-IDA; 1 L WZ-11947-30 Mfr # KN40650410
COMPARE
Cobalt & Iminodiacetic acid resin 1000
Call For Price
Bio-Works WorkBeads 40 Zn High Capacity IMAC Medium for His-Tagged Proteins, Zn-IDA; 25 mL WZ-11947-31 Mfr # KN40650501
COMPARE
Zinc & Iminodiacetic acid resin 25
Call For Price
Bio-Works WorkBeads 40 Zn High Capacity IMAC Medium for His-Tagged Proteins, Zn-IDA; 150 mL WZ-11947-32 Mfr # KN40650503
COMPARE
Zinc & Iminodiacetic acid resin 150
Call For Price
Showing 1 - 20 of 33