Wire Mesh Overscreens for Sanitary Strainers
Protect products and expensive process equipment from coarse or fine particulates
  • Mesh overscreens are available in a variety of mesh sizes to meet your specific application
  • Overscreens are hemmed smooth with no stray wire ends
  • Overscreens fit both in-line and side-inlet sanitary strainer assemblies
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.
£109.20 - £151.20GBP / Each
£131.04 - £181.44GBP (incl VAT)
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Mesh Size Micron Nominal (micron)
Availability Pricing
COMPARE
20 x 20 864
£114.45GBP / Each
£137.34GBP (incl VAT)
COMPARE
30 x 30 516
£109.20GBP / Each
£131.04GBP (incl VAT)
COMPARE
40 x 40 381
£109.20GBP / Each
£131.04GBP (incl VAT)
COMPARE
60 x 60 234
£109.20GBP / Each
£131.04GBP (incl VAT)
COMPARE
80 x 80 178
£109.20GBP / Each
£131.04GBP (incl VAT)
COMPARE
100 x 100 140
£143.85GBP / Each
£172.62GBP (incl VAT)
COMPARE
150 x 150 104
£151.20GBP / Each
£181.44GBP (incl VAT)
Showing 1- 7 of 7