Wika 4" Stainless Steel Test Gauges
Ideal for applications demanding high precision and consistent results
  • Accuracy is ±0.25% full scale (ANSI/ASME Grade 3A)
  • Knife-edge pointer tip and mirrored band eliminate parallax reading errors.
  • The 6" dial gauges feature a low-friction, wear-resistant nickel-silver movement that delivers a long service life.
Gauge construction includes a 316 stainless steel (SS) socket; a 316 SS Bourdon tube for the 4" dial gauges or a copper alloy tube for the 6" dial gauges; a 316 SS fitting; a 304 SS case; and a glass lens. All gauges have a 1/4" NPT(M) process connection. The 4" dial gauges include a zippered, soft nylon carrying case.
£223.41 - £234.55GBP / Each
£268.09 - £281.46GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Wika 332.54 4 4" Test Gauge, 30 to 0" Hg, 316 Stainless Steel WZ-68043-00 Mfr # 4220013
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 14.735 0.1\" Hg
£223.41GBP / Each
£268.09GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 15 psi, 316 Stainless Steel WZ-68043-02 Mfr # 4220021
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 15 0.05 psi
£227.96GBP / Each
£273.55GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 30 psi, 316 Stainless Steel WZ-68043-04 Mfr # 4220030
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 30 0.1 psi
£234.55GBP / Each
£281.46GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 60 psi, 316 Stainless Steel WZ-68043-06 Mfr # 4220048
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 60 0.2 psi
£226.37GBP / Each
£271.64GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 100 psi, 316 Stainless Steel WZ-68043-08 Mfr # 4220056
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 100 0.5 psi
£224.32GBP / Each
£269.18GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 200 psi, 316 Stainless Steel WZ-68043-10 Mfr # 4220072
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 200 0.5 psi
£227.96GBP / Each
£273.55GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 400 psi, 316 Stainless Steel WZ-68043-12 Mfr # 4220099
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 400 1 psi
£225.46GBP / Each
£270.55GBP (incl VAT)
Wika 332.54 4 4" Test Gauge, 0 to 600 psi, 316 Stainless Steel WZ-68043-14 Mfr # 4220102
COMPARE
±0.25% full-scale (ANSI/ASME B40.1 Grade 4A) 600 2 psi
£227.96GBP / Each
£273.55GBP (incl VAT)
Showing 1- 8 of 8