Wika Low Pressure Capsule Gauges - Type 611.10
Accurately measuring pressures as low as 10 psi
  • Ideal for gaseous, dry, and non-aggressive media
  • Front zero adjustment
These low pressure gauges are extremely sensitive and highly accurate. The copper alloy capsule is designed to measure pressure and vacuum of gaseous media from as low as 15" H2O to 240" H2O (10 psi). An adjustment screw is located on the dial to re-zero the pointer if necessary. The finely polished nickel silver pinion gear and shaft of the movement ensure repeatable accuracy.
£68.55GBP / Each
£82.26GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (in WC) Description
Availability Pricing
Wika 9851933 Low Pressure Capsule Gauge, 0 to 5 psi, 2.5" Dual Scale, 1/4" NPT(M) WZ-68434-55 Mfr # 9851933
COMPARE
±1.5% full-scale 138.54 2.5\" Dual Scale Vacuum Gauge, 0 to 5 psi
£68.55GBP / Each
£82.26GBP (incl VAT)