Chemware Wide-Mouth Wash Bottles, PFA
Ideal for all kinds of liquids
  • Resistant to most chemicals
  • Break resistant
  • Easy to fill and clean
Transparent, break-resistant bottles are inert to virtually all chemicals—use to dispense all kinds of liquids. Bottles are ideal for ultra-pure environmental and analytical work. Wide mouth makes bottles easy to fill. Nonstick and easy to clean.
£155.99 - £537.59GBP / Each
£187.19 - £645.11GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material
Availability Pricing
Chemware D1069739 Wash Bottle, Wide-Mouth, PFA, 100 mL, 1/Pk WZ-62305-00 Mfr # D1069739
COMPARE
100 Perfluoroalkoxy (PFA)
£155.99GBP / Each
£187.19GBP (incl VAT)
Chemware D1069740 Wash Bottle, Wide-Mouth, PFA, 250 mL, 1/Pk WZ-62305-10 Mfr # D1069740
COMPARE
250 Perfluoroalkoxy (PFA)
£201.11GBP / Each
£241.33GBP (incl VAT)
Chemware D1069741 Wash Bottle, Wide-Mouth, PFA, 500 mL, 1/Pk WZ-62305-20 Mfr # D1069741
COMPARE
500 Perfluoroalkoxy (PFA)
£319.56GBP / Each
£383.47GBP (incl VAT)
Chemware D1069742 Wash Bottle, Wide-Mouth, PFA, 1 L, 1/Pk WZ-62305-30 Mfr # D1069742
COMPARE
1000 Perfluoroalkoxy (PFA)
£537.59GBP / Each
£645.11GBP (incl VAT)