Wheaton Open Water Grab Samplers
Simple pull-ring operation lets you draw samples at desired depth
  • Break down to 6-ft sections for easy transport
Inert PP head with high-strength aluminum pole assembly. Samplers include a 1000-mL glass bottle and cap—accept any GPI 38-430 screw thread bottle.
£69.03GBP / Each
£82.84GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material
Availability Pricing
DWK Life Sciences (Kimble) 990477 Replacement Bottle/Cap for Open Water Grab Sampler WZ-99152-50 Mfr # 990477
COMPARE
0.4 Glass
£69.03GBP / Each
£82.84GBP (incl VAT)