WHEATON® Battery-Operated Crimping and Decapping Tools
Reduce crimping and decapping time
  • Ergonomically designed to reduce strain
  • Use with flip-off caps
Quickly and easily crimp and decap laboratory vials and seals with aluminum or two-part aluminum/steel caps. These battery-powered crimping tools come with a powerful motor with a fast cycle. Long lasting battery provides up to 800 crimps per charge and even lets you operate while charging. The electronically adjustable cycle is accurate and reproducible and can be adjusted with control pad on top of the unit. Use with optional tool stand to reduce hand fatigue.
£1,925.86GBP / Each
£2,311.03GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Cap Diameter (mm)
Availability Pricing
DWK Life Sciences (Wheaton) W225714 Vial Decapper, 13 mm, with Rechargeable Battery WZ-98800-06 Mfr # W225814
COMPARE
Battery-powered decapper 13
£1,925.86GBP / Each
£2,311.03GBP (incl VAT)