This product is now discontinued, View Related Products

Water Level Loggers

Datalogging capabilities let you remotely monitor and record water levels
  • Ideal for use with ground water, surface water, storm water, and sewers
  • Temperature- and barometric-pressure compensated readings with vented cable
These highly accurate waterlevel loggers can record over 81,000 readings. Program the logger to store data with one of four programs: fast (10 measurements per second), programmable interval (1 second to multiple years), logarithmic, and exception. Cable lengths up to 500 feet and serial communications models available; contact our Application Specialists for details.

Units include Windows®-based software for easy option setting, stored data access, and data download into a standard spreadsheet. The software also provides additional features such as real-time monitoring, measurement interval with start and stops, engineering unit selections, station ID, and sensor calibration. USB connections provide convenient connection for downloads without having to remove the sensor from the well.

Weatherproof loggers easily mount to a 2'' wellhead with standard hardware. The internal 9 V batteries power the logger and sensor for up to a year, depending on the recording interval. A third battery retains data even if the other two batteries fail.
Call For Price

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Global Water WL16U-003-003 Water-Level Logger 3 Foot Range, 3 Foot Cable WZ-32922-50 Mfr # WL16U-003-003
COMPARE
Water-Level Logger 3 Foot Range, 3 Foot Cable
Call For Price
Global Water WL16U-003-025 Water-Level Logger, 3 foot range with 25 foot cable WZ-32922-52 Mfr # WL16U-003-025
COMPARE
Water-Level Logger, 3 foot range with 25 foot cable
Call For Price
Global Water WL16U-015-025 Water-Level Logger, 15 foot range with 25 foot cable WZ-32922-54 Mfr # WL16U-015-025
COMPARE
Water-Level Logger, 15 foot range with 25 foot cable
Call For Price
Global Water WL16U-015-050 Water-Level Logger, 15 foot range with 50 foot cable WZ-32922-56 Mfr # WL16U-015-050
COMPARE
Water-Level Logger, 15 foot range with 50 foot cable
Call For Price
Global Water WL16U-030-025 Water-Level Logger, 30 foot range with 25 foot cable WZ-32922-58 Mfr # WL16U-030-025
COMPARE
Water-Level Logger, 30 foot range with 25 foot cable
Call For Price
Global Water WL16U-030-050 Water-Level Logger, 30 foot range with 50 foot cable WZ-32922-60 Mfr # WL16U-030-050
COMPARE
Water-Level Logger, 30 foot range with 50 foot cable
Call For Price
Global Water WL16U-060-075 Water-Level Logger, 60 foot range with 75 foot cable WZ-32922-62 Mfr # WL16U-060-075
COMPARE
Water-Level Logger, 60 foot range with 75 foot cable
Call For Price
Global Water WL16U-060-100 Water-Level Logger, 60 foot range with 100 foot cable WZ-32922-64 Mfr # WL16U-060-100
COMPARE
Water-Level Logger, 60 foot range with 100 foot cable
Call For Price
Global Water WL16U-120-125 Water-Level Logger, 120 foot range with 125 foot cable WZ-32922-66 Mfr # WL16U-120-125
COMPARE
Water-Level Logger, 120 foot range with 125 foot cable
Call For Price
Global Water WL16U-120-150 Water-Level Logger, 120 foot range with 150 foot cable WZ-32922-68 Mfr # WL16U-120-150
COMPARE
Water-Level Logger, 120 foot range with 150 foot cable
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat