This product is now discontinued, View Related Products
Walker Portable Precision Gaussmeter
Package includes CC-4D Carrying Case, 6' Probe Cable, Batteries
and NIST Calibration Certificate.
FEATURES
High accuracy and resolution
Measure DC and AC fields; True RMS readings to 20 kHz with AC probes
Range extendable to 150 kG with select probes
Analog Output
High impact foam-lined plastic carrying case
Operates up to one year with standard “AA” alkaline batteries
APPLICATIONS
Measure residual fields
Analyze magnetic circuits and components
Classify magnets
Measure absolute and differential fields
Plot field uniformity
Measure stray and leakage fields
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Resistance Accuracy Resistance Range Resistance Resolution
Availability Pricing
Laboratorio Electrofisco 60-B411756C Portable Gaussmeter w/ Transverse Probe WZ-20001-61 Mfr # 60-B411756C
COMPARE
Call For Price
Laboratorio Electrofisco 60-B411756H Portable Gaussmeter w/ Axial Probe WZ-20001-62 Mfr # 60-B411756H