This product is now discontinued, View Related Products

VOC-TRAQ USB PID Toxic Gas Detectors

PID gas detector with the convenience of a USB output
Ideal for indoor air quality studies, safety engineers, maintenance managers, leak detection applications, and more! Software allows you to set an alarm level to a specific ppm value and enable/disable audible alarms. Red blinking LED indicates an alarm condition has been reached; yellow blinking LED indicates an alarm condition that has passed. Operate while connected to a computer capturing data through the USB port or use remotely, storing up to 36,000 readings in internal memory by using the optional rechargeable battery pack.
Call For Price

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Datalogging Gases Measured Description
Availability Pricing
Baseline Mocon 043-207CP USB PID Toxic Gas Detector, 0.1 to 100 ppm (benzene), 9.6 eV lamp energy WZ-86262-30 Mfr # 043-207CP
COMPARE
USB PID Toxic Gas Detector, 0.1 to 100 ppm (benzene), 9.6 eV lamp energy
Call For Price
Baseline Mocon VOC-TRAQ USB PID Toxic Gas Detector; 1 to 1000 ppm Isobutylene WZ-86262-31 Mfr # 043-209CP
COMPARE
Yes PID USB PID Toxic Gas Detector; 1 to 1000 ppm Isobutylene
Call For Price
Baseline Mocon VOC-TRAQ USB PID Toxic Gas Detector; 10 to 10,000 ppm Isobutylene WZ-86262-32 Mfr # 043-208CP
COMPARE
Yes Isobutylene USB PID Toxic Gas Detector; 10 to 10,000 ppm Isobutylene
Call For Price
Baseline Mocon VOC-TRAQ USB PID Toxic Gas Detector; 0.02 to 20 ppm Isobutylene WZ-86262-33 Mfr # 042-963CP
COMPARE
Yes PID USB PID Toxic Gas Detector; 0.02 to 20 ppm Isobutylene
Call For Price
Baseline Mocon VOC-TRAQ USB PID Toxic Gas Detector; 0.2 to 200 ppm Isobutylene WZ-86262-34 Mfr # 042-962CP
COMPARE
Yes PID USB PID Toxic Gas Detector; 0.2 to 200 ppm Isobutylene
Call For Price
Baseline Mocon VOC-TRAQ USB PID Toxic Gas Detector; 2 to 2000 ppm Isobutylene WZ-86262-35 Mfr # 042-961CP
COMPARE
Yes PID USB PID Toxic Gas Detector; 2 to 2000 ppm Isobutylene
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat