This product is now discontinued, View Related Products
ViscoDispo Disposable Viscous Media Samplers
Perfect for taking samples of high-viscosity fluids
  • Ergonomic handle for easy sampling
  • Cleanroom manufactured
  • Individually packaged
Ergonomically designed handle, a sturdy draw bar, and a large opening make it easy to sample highly viscous liquids up to 100,000 mPas. These single-use HDPE samplers are manufactured in a cleanroom and individually packaged to prevent contamination ensuring accurate sampling results.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon ViscoDispo Disposable Viscous Media Samplers, 50 cm L, 100 mL; Pk of 20 WZ-33687-58 Mfr # 853933-3320
COMPARE
19.625 ViscoDispo Disposable Viscous Media Samplers, 50 cm L, 100 mL; Pk of 20
Call For Price
Dynalon ViscoDispo Disposable Viscous Media Samplers, Sterile, 50 cm L, 100 mL; Pk of 20 WZ-33687-59 Mfr # 853933-3321
COMPARE
19.625 ViscoDispo Disposable Viscous Media Samplers, Sterile, 50 cm L, 100 mL; Pk of 20
Call For Price
Dynalon ViscoDispo Disposable Viscous Media Samplers, 100 cm L, 200 mL; Pk of 20 WZ-33687-60 Mfr # 853933-3330
COMPARE
39.375 ViscoDispo Disposable Viscous Media Samplers, 100 cm L, 200 mL; Pk of 20
Call For Price
Dynalon ViscoDispo Disposable Viscous Media Samplers, Sterile, 100 cm L, 200 mL; Pk of 20 WZ-33687-61 Mfr # 853933-3331
COMPARE
39.375 ViscoDispo Disposable Viscous Media Samplers, Sterile, 100 cm L, 200 mL; Pk of 20