This product is now discontinued, View Related Products

Venting Filters

Manufactured under strict cleanroom conditions and totally biosafe
Filters feature a polypropylene housing and a 0.2 µm PTFE membrane allowing easy scale-up within the range based on vessel capacity and flow rate required. These filters are ideal for venting and isolation applications to protect vessel contents from external contaminants in biotechnology, pharmaceutical, and food and beverage industries. Filters are also used to protect the environment from contaminants within the vessel.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Effective Filtration Area (cm²)
Availability Pricing
ZenPure Venting filter disc, 25mm, 50/pk (Female luer/Male luer) WZ-49700-60 Mfr # D25CF020LFLS-25
COMPARE
4.6
Call For Price
ZenPure Venting filter disc, 50mm, 10/pk (1/4" to 3/8" stepped hosebarb) WZ-49700-62 Mfr # D50CF0202H2H-10
COMPARE
15.9
Call For Price
ZenPure Venting filter capsule, 500 cm<sup>2</sup> WZ-49700-64 Mfr # MKPF020D2H2H-N
COMPARE
500
Call For Price
ZenPure Venting filter capsule, 1000 cm<sup>2</sup> WZ-49700-66 Mfr # SKLF020S3H3H-N
COMPARE
1250
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat