This product is now discontinued, View Related Products

VELP® FIWE Raw Fiber Extractors

Rapid analysis in just two hours
  • Reliable results with high ±1% reproducibility
  • No sample loss—same crucibles used for weighing, drying, and washing
  • Complete system includes hot plate for preheating the reagents
These raw fiber extractors feature rapid analysis, reliable results, and high reproducibility. They are ideal for total raw fiber extraction using Weende technique, neutral and acid detergent fiber determination (NDF and ADF according to Van Soest), acid detergent lignin extraction (ADL according to Van Soest), and fiber fraction analysis such as cellulose, hemicellulose and pectin.

Perform single or multiple extractions including boiling, rinsing, and filtration. The extremely efficient heating element speeds up the analysis, saving you time.
Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Number of Positions Power (watts) Description
Availability Pricing
Velp FIWE3 Raw Fiber Extractor, 3 positions; 115V/60Hz WZ-86580-54 Mfr # SA30540201
COMPARE
3 900 Raw Fiber Extractor, 3 positions; 115V/60Hz
Call For Price
Velp FIWE 6 Raw Fiber Extractor, 6 positions; 115V/60Hz WZ-86580-55 Mfr # SA30540200
COMPARE
6 1200 Raw Fiber Extractor, 6 positions; 115V/60Hz
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat