This product is now discontinued, View Related Products
Velp AREX Round-Top Ceramic-Coated Stirring Hot Plate
Ceramic-coated top plate combines temperature uniformity with durability
  • Aluminum plate ensures optimal heat transfer while ceramic coating promotes excellent chemical resistance
  • Innovative low-profile design for maximum comfort
This hot plate stirrer ensures precise thermoregulation of the plate as well as a high degree of reliability and safety. The control panel is separated from the heating plate, and leaks or spills are channeled away from the controls by the dedicated run-off groove. The pressure die-cast structure ensures that accidental liquid spills cannot reach the internal parts of the unit. The raised front panel features easy-to-use temperature and speed controls and the ceramic-coated heating plate provides easy cleaning and resistance to scratches and surface abrasions. For maximum control, connect the optional Vertex temperature controller for direct temperature control of the liquid (sold separately).
£636.99GBP / Each
£764.39GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Velp F20510413 AREX Ceramic Coated Stirring Hot Plate, Round Top, 115 VAC. WZ-86579-50 Mfr # F20510413
COMPARE
115
£636.99GBP / Each
£764.39GBP (incl VAT)
Velp F20500413 AREX Ceramic Coated Stirring Hot Plate, Round Top, 230 VAC, 50/60 Hz. WZ-86579-51 Mfr # F20500413
COMPARE
230
£636.99GBP / Each
£764.39GBP (incl VAT)
Velp F20530163 AREX Ceramic Coated Stirring Hot Plate, Round Top, 230 VAC, 60 Hz. WZ-86579-52 Mfr # F20530163
COMPARE
230