Velp AREX Digital Round-Top Ceramic-Coated Stirring Hot Plates
Counter-reaction technology (SpeedServo™) ensures a constant speed even if sample viscosity changes
  • Aluminum alloy top provides uniform heat distribution over the entire surface
  • Protective ceramic coating for easy cleaning and excellent resistance to chemicals, scratches, and surface abrasions
These stirring hot plates are designed for a high degree of reliability, safety, and usefulness. The control panel is separated from the hot plate by a dedicated run-off groove designed to channel away any leaks or spills increasing the safety and durability of the unit. The digital displays make it simple to set and monitor both temperature and stirring speed. A "hot plate warning" icon stays activated until the plate temperature cools below 122°F (50°C). Further safety features include a user- programmed maximum temperature setting, preventing the stirrer from heating beyond the desired temperature while also preventing unintentional heat up.
£1,231.51GBP / Each
£1,477.81GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Velp SA20510411 AREX Digital Ceramic Coated Stirring Hot Plate, Round Top; 115 VAC WZ-86579-53 Mfr # SA20510411
COMPARE
115
Call For Price
Velp SA20500411 AREX Digital Ceramic Coated Stirring Hot Plate, Round Top; 230 VAC WZ-86579-54 Mfr # SA20500411
COMPARE
230
Call For Price
Velp SB20510410 AREX Digital Pro Ceramic Coated Stirring Hot Plate, Round Top; 115 VAC WZ-86579-55 Mfr # SB20510410
COMPARE
115
£1,231.51GBP / Each
£1,477.81GBP (incl VAT)
Velp SB20500410 AREX Digital Pro Ceramic Coated Stirring Hot Plate, Round Top; 230 VAC WZ-86579-56 Mfr # SB20500410
COMPARE
230
£1,231.51GBP / Each
£1,477.81GBP (incl VAT)