This product is now discontinued, View Related Products
Velp ARE Stirring Hot Plate
Optimal heat transfer with uniform distribution
  • Aluminum top combines excellent conductivity and temperature uniformity with good chemical resistance
  • low profile, curved design for maximum comfort
Excellent for both industrial and laboratory applications, the ARE stirring hot plate is designed to last. The aluminum alloy top plate ensures accurate heating and has a protective coating providing excellent chemical resistance. Designed with an elevated control panel separated from the plate, a dedicated run-off groove channels spills away from the controls for added safety and durability.
£540.98GBP / Each
£649.18GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Velp F20510162 ARE Hot Plate Stirrer, Round Aluminum Top, 115 VAC. WZ-86579-37 Mfr # F20510162
COMPARE
115
£540.98GBP / Each
£649.18GBP (incl VAT)
Velp F20500162 ARE Hot Plate Stirrer, Round Aluminum Top, 230 VAC, 50/60 Hz.. WZ-86579-38 Mfr # F20500162
COMPARE
230
£540.98GBP / Each
£649.18GBP (incl VAT)
Velp F20530162 ARE Hot Plate Stirrer, Round Aluminum Top, 230 VAC, 60 Hz.. WZ-86579-39 Mfr # F20530162
COMPARE
230