This product is now discontinued, View Related Products
Vector Laboratories ImmPRESS™ Polymer Kits
Provides highly sensitive, one-step detection
  • Shorter assay time
  • Simplified multiple labeling
The ImmPRESS polymer kits offer a sensitive, ready-to-use, one-step detection system for immunohistochemistry and immunocytochemistry staining. The kits feature micropolymers of highly active enzymes (peroxidase or alkaline phosphatase) attached to purified secondary antibodies, producing reagents with outstanding sensitivity and low background The micropolymers of the reagent limit steric interference and provide enhanced accessibility to the target. The result is crisp, strong staining of antibody targets, especially nuclear and membrane antigens.
The reagents included in all the kits come prediluted in ready-to-use dropper bottles along with prediluted blocking serum. No mixing or titering is necessary to obtain optimal tissue section staining.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Capacity (Number of slides) Description
Availability Pricing
Vector Laboratories ImmPRESS AP Anti-Rabbit IgG Reagent, made in horse, 50 ml WZ-93951-26 Mfr # MP-5401-50
COMPARE
50 250 to 500 AP Anti-Rabbit IgG Reagent, made in horse, 50 ml
Call For Price
Vector Laboratories ImmPRESS AP Anti-Mouse IgG Reagent, made in horse, 50 ml WZ-93951-28 Mfr # MP-5402-50
COMPARE
50 250 to 500 AP Anti-Mouse IgG Reagent, made in horse, 50 ml
Call For Price
Vector Laboratories ImmPRESS VR Polymer HRP Anti-Mouse IgG Reagent, 15 ml WZ-93951-30 Mfr # MP-6402-15
COMPARE
15 75 to 150 VR Polymer HRP Anti-Mouse IgG Reagent, 15 ml
Call For Price
Vector Laboratories ImmPRESS Universal Antibody Kit, 15 ml WZ-93951-58 Mfr # MP-7500-15
COMPARE
15 75 to 150 Universal Antibody Kit, 15 ml
Call For Price
Vector Laboratories M.O.M. ImmPRESS HRP Ig Reagent Kit WZ-93951-65 Mfr # MPX-2402-15
COMPARE
15 75 to 150 HRP Ig Reagent Kit