Vaisala DRYCAP Dew Point Transmitter
Ideal choice for compressed air lines
  • Rapid response time with long-term stability
  • NIST traceable
Vaisala DRYCAP DMT-series transmitter provides reliable and stable measurements for low dew point industrial dryer applications. The sensor is immune to particulate contamination, water condensation, oil vapor, and most chemicals. Because the sensor withstands condensation, it offers unmatched performance in low dew point applications that experience process water spikes. The housing is IP65 (NEMA 4) rated and provides protection against dust, dirt and splashed water. Due to its compact size it can quickly and easily be installed in tight spaces and directly into a system with a maximum pressure of 20 bar. The transmitter can be field checked using Vaisala handheld meters
£1,562.40 - £1,745.70GBP / Each
£1,874.88 - £2,094.84GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Max RH Range (%) Mounting Type
Availability Pricing
Vaisala DMT242A1D1A0B Industrial dew Point Transmitter with Plastic Housing WZ-37803-43 Mfr # DMT143N1C1A1D3A0ASX
COMPARE
£1,562.40GBP / Each
£1,874.88GBP (incl VAT)
Vaisala DMT143N1C1A1A3A0ASX Industrial dew Point Transmitter with Metal Housing WZ-37804-43 Mfr # DMT143N1C1A1A3A0ASX
COMPARE
£1,745.70GBP / Each
£2,094.84GBP (incl VAT)