Two-Way PVC Ball Valves, Female NPT
Create a secure pipe threaded connection
  • Constructed of durable PVC
Control flow with the 90°> rotating handle. Valves have seals made of Viton® fluoroelastomer and seats made of PTFE. Maximum pressure is 150 psi at 32°F
(10.3 bar at 0°C); maximum temperature is 140°F
(60°C at 1.7 bar).
£23.92 - £36.81GBP / Each
£28.70 - £44.17GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
COMPARE
8 1/2" NPT(F) 1/2" NPT(F)
£23.92GBP / Each
£28.70GBP (incl VAT)
COMPARE
15 3/4" NPT(F) 3/4" NPT(F)
£29.22GBP / Each
£35.06GBP (incl VAT)
COMPARE
29 1" NPT(F) 1" NPT(F)
£36.81GBP / Each
£44.17GBP (incl VAT)