GFS Chemicals Turbidity Standard Solutions
Ensure accurate readings by calibrating your turbidimeter
  • EPA-approved polymeric standards outlast Formazin standards
  • Suspended polymeric particles virtually never settle or clump
Primary Standard Solutions can be used to calibrate nephelometers or turbidimeters. Solutions are guaranteed to maintain NTU values within ±1% for one year. Available in one-liter bottles.
£111.73 - £223.46GBP / Each
£134.08 - £268.15GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
NTU Value
Availability Pricing
GFS Chemicals Turbidity Primary Standard Solution, 1.0 NTU, 1 L WZ-08391-03 Mfr # 85034
COMPARE
1.0
£205.67GBP / Each
£246.80GBP (incl VAT)
GFS Chemicals Turbidity Standard, 10.0 NTU, 1-L bottle WZ-08391-04 Mfr # 85063
COMPARE
10
£200.97GBP / Each
£241.16GBP (incl VAT)
GFS Chemicals Turbidity Standard, 40.0 NTU, 1-L bottle WZ-08391-05 Mfr # 85075
COMPARE
40
£201.11GBP / Each
£241.33GBP (incl VAT)
GFS Chemicals Turbidity Primary Standard Solution, 0.2 NTU, 1 L WZ-08391-08 Mfr # 85014
COMPARE
0.2
Call For Price
GFS Chemicals Turbidity Primary Standard Solution, 0.5 NTU, 1 L WZ-08391-09 Mfr # 85023
COMPARE
0.5
£223.46GBP / Each
£268.15GBP (incl VAT)
GFS Chemicals Turbidity Primary Standard Solution, 0 NTU, 1 L WZ-08391-19 Mfr # 85005
COMPARE
0.0
£111.73GBP / Each
£134.08GBP (incl VAT)