This product is now discontinued, View Related Products
TriForest Wide-Mouth Bottles, PC
Engineered for ultra-centrifugation and culturing
  • Culture microbes, plants, or algae—large surface area to volume ratio
  • Polycarbonate (PC) is a safer, shatterproof, alternative to glass
  • Leakproof seals (included) secure samples and prevent aerosol escape
  • Bottle and cap will both withstand reuse and multiple autoclave cycles
  • Molded-in graduations make reference fills easy
Use these clear, shatterproof, polycarbonate bottles to store critical fluids or as ultracentrifuge flasks—bottles withstand maximum speeds without leakage.
Autoclavable
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material
Availability Pricing
TriForest WPC0180 Wide-Mouth Bottle, 180 mL, PC, Screw, pack of 12, 144/CS WZ-06542-31 Mfr # WPC0180
COMPARE
180 Polycarbonate(PC)
Call For Price
TriForest WPC0300 Wide-Mouth Bottle, 300 mL, PC, Screw, pack of 24, 96/CS WZ-06542-32 Mfr # WPC0300
COMPARE
300 Polycarbonate(PC)
Call For Price
TriForest WPC0400 Wide-Mouth Bottle, 400 mL, PC, Screw, pack of 12, 72/CS WZ-06542-33 Mfr # WPC0400
COMPARE
400 Polycarbonate(PC)
Call For Price
TriForest WPC0500 Wide-Mouth Bottle, 500 mL, PC, Screw, pack of 12, 72/CS WZ-06542-34 Mfr # WPC0500
COMPARE
500 Polycarbonate(PC)
Call For Price
TriForest WPC1000 Wide-Mouth Bottle, 1000 mL, PC, Screw, pack of 12, 48/CS WZ-06542-35 Mfr # WPC1000
COMPARE
1000 Polycarbonate(PC)