This product is now discontinued, View Related Products
TriForest Round Media Bottles
Durable leakproof design for safe sample storage
  • Printed graduations for accurate measurement
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom that is ISO 9001 compliant
  • Autoclavable bottle and PP cap withstand repeated cycles
Made of either polycarbonate (PC) or polypropylene (PP), these bottles are nonleaching and shatterproof, being a safer alternative to glass in storing media, buffers, reagents, and other aqueous solutions. Optically superior transparent bottles and blue caps make visual identification of samples quick and easy.
Bottles are provided nonsterile in a shrink-wrapped tray. The 500- and 1000-mL capacities have 12 bottles per tray and the 300-mL capacity has 24 bottles per tray.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
TriForest Boston Round Media Bottles, 300 mL, PC; 96/Cs WZ-06542-26 Mfr # RPC0300
COMPARE
300 Polycarbonate(PC) 5 51/64
Call For Price
TriForest Boston Round Media Bottles, 500 mL, PC; 72/Cs WZ-06542-27 Mfr # RPC0500
COMPARE
500 Polycarbonate(PC) 7 1/10
Call For Price
TriForest Boston Round Media Bottles, 1000 mL, PC; 48/Cs WZ-06542-28 Mfr # RPC1000
COMPARE
1000 Polycarbonate(PC) 8 45/64
Call For Price
TriForest Boston Round Media Bottles, 500 mL, PP; 72/Cs WZ-06542-29 Mfr # RPP0500
COMPARE
500 Polypropylene(PP) 6 51/64
Call For Price
TriForest Boston Round Media Bottles, 1000 mL, PP; 48/Cs WZ-06542-30 Mfr # RPP1000
COMPARE
1000 Polypropylene(PP) 8 2/5