tracrack® Cold Storage System
Thaw what you need and keep the rest frozen
  • Removable tube racks let you take a subset of samples and leave the rest in storage
  • Reduces cross-contamination when moving samples between storage and working racks
  • Durable polypropylene safely stores samples down to -80°C
These cold storage boxes feature removable rows of tube racks, so you can pull out one row of samples at a time and leave the rest in the freezer. Each row can be used as a working rack, preventing cross-contamination. Open hinged-lid tubes do interfere with each other and alphanumeric well identification helps reduce errors.
£25.86GBP / Each
£31.03GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Heathrow Scientific tracrack® Cold Storage System, 1.5 / 2.0 mL tubes, 1/ea WZ-06696-25 Mfr # HS120356
COMPARE
Cold Storage System, 1.5 / 2.0 mL tubes, 1/ea
£25.86GBP / Each
£31.03GBP (incl VAT)