Monarch Instrument Track-It DC Voltage and Current Data Loggers
Rugged overall cover protects logger in dirty environments
  • ±0.5% accuracy
  • Records 500mVDC, 1VDC, 5VDC, 10VDC, or 20mA
  • Capable of recording temperature
  • Optional three-year battery for long-term recording
Use these data loggers for various DC voltage and current applications, such as monitoring industrial processes, productivity checks, and troubleshooting. Each data logger is a battery powered stand alone compact data logger that records up to 64,000 samples each of DC voltage and current data. Record temperature readings if you choose by not connecting any modules. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. You can choose from a model with a standard battery or one with a long-life battery.
Track-It™ software is a features-packed software package that is included with every data logger. It allows for easy setup, retrieval, interpretation and export of the recorded data. You can view a numerical data table or graphical representation of DC voltage/current readings relative to time. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger, USB interface cable sold separately. The software can also turn your PC into a real-time data acquisition system by allowing you to stream voltage or current data as it occurs directly to your PC. Use the one click export function to automatically export stored data into an Excel® spreadsheet.
£144.10 - £185.00GBP / Each
£172.92 - £222.00GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
DC Current Accuracy (ADC) Max Current (ADC) Max Temperature (° F)
Availability Pricing
Monarch 5396-0511 Voltage Data Logger, LCD Display; Standard Battery WZ-30003-05 Mfr # 5396-0511
COMPARE
£144.10GBP / Each
£172.92GBP (incl VAT)
Monarch TI-1V/DIS/STDB Analog 1 V Track It Logger with display, 1 V module and standard battery WZ-30003-07 Mfr # 5396-0512
COMPARE
£144.10GBP / Each
£172.92GBP (incl VAT)
Monarch 5396-0514 Analog 10 V Track It Logger with display, 10 V module and standard battery WZ-30003-11 Mfr # 5396-0514
COMPARE
£144.10GBP / Each
£172.92GBP (incl VAT)
Monarch Instrument TI-10V/DIS/LLB Analog 10 V Track It Logger with display, 10 V module and long-life battery WZ-30003-12 Mfr # 5396-0524
COMPARE
£185.00GBP / Each
£222.00GBP (incl VAT)
Monarch 5396-0515 Current Data Logger, LCD Display; Standard Battery WZ-30003-13 Mfr # 5396-0515
COMPARE
£144.10GBP / Each
£172.92GBP (incl VAT)
Monarch 5396-0525 Current Data Logger, LCD Display; Long-Life Battery WZ-30003-14 Mfr # 5396-0525
COMPARE
£175.46GBP / Each
£210.55GBP (incl VAT)