Traceable® Big-Digit 4-Alarm Digital Thermometers with Calibration
Four different alarm configurations
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Jumbo digital display is easy to read from a distance
  • Remote, heat-resistant probe
This thermometer has two temperature sensors—a built-in air sensor and an independent external probe sensor with 3 1/2 foot cable. The external probe can be used to monitor temperatures of liquids, air/gas, and semisolids.
Alarm configurations can be set up for four conditions: temperature above a setting, below a setting, between two settings, or outside two settings. A green or red light will blink and an audible alarm will sound when one of these conditions is detected. The alarm will sound every minute for 5 seconds until the temperature returns to a non-alarm condition or until the alarm is turned off.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
£35.66GBP / Each
£42.79GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
158 -58
£35.66GBP / Each
£42.79GBP (incl VAT)