Torque and Angle Electronic Torque Wrenches
Digital torque and angle measurements in one convenient wrench
  • ±2% accuracy CW of indicated value
  • Gyro chip senses 360° of movement, and is accurate to within ±1°
  • Handle vibrates when target torque is reached; almost impossible to over torque
  • 3-step LED visual signal system:
  • Yellow = approaching target torque
  • Green = target torque reached
  • Red = over torque indicator
  • Ergonomic non-slip cushion grip
  • 15% Flex ratchet head and push-button socket release head
  • Low maintenance — change batteries at 80 hours
  • Meets or exceeds ASME B107.28 - 2005
These electronic torque and angle wrenches incorporate the latest technology into a simple-to-use digital wrench. Specifically designed to meet the requirements where torque plus angle is required and features three units of torque — ft. lbs., in.lbs., and Nm, plus an angle mode for additional flexibility.
Wrenches allow you to apply a specific torque load, followed by the angle specification, all in one convenient wrench. Additional features include an easy-to-read digital display, LED light bar indicator, audible buzzer, tactile vibrating handle, and separate function that allows the wrench to be used as a stand-alone digital torque wrench.
£1,364.73GBP / Each
£1,637.68GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m)
Availability Pricing
J. H. Williams Tool Group 2503TAA-CDI Torque and Angle Electronic Torque Wrench, 25 to 250 ft. lb. WZ-25301-14 Mfr # 2503EFRMH
COMPARE
3000 338
£1,364.73GBP / Each
£1,637.68GBP (incl VAT)