Thermo Scientific™ TRACE TR-WAX (Polyethylene glycol [PEG]) GC Columns
Low bleed provides reproducible and accurate results
  • Suitable for Polyethylene glycol (PEG) phase
Thermo Scientific TRACE GC columns have been tested and found to provide less than half the bleed of competitive capillary columns. Low bleeding means fewer problems with low sensitivity, contamination of the detector, lower temperature limits, and shorter life spans. These columns have been manufactured with improved bonding to be extremely robust, minimizing the risk of damage due to contaminated carrier gases. Improved column-to-column reproducibility guarantees that your column will work with the same high standard as those purchased years from now. Each column goes through rigorous testing and is delivered with a certificate of traceability to guarantee the performance.
TRACE TR-WAX Columns are general purpose high polarity columns that are ideal for all polar compounds and provide excellent peak shape even when working with short-chain alcohols.
£881.76GBP / Each
£1,058.11GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (m) Film Thickness (µm)
Availability Pricing
Thermo Scientific TRACE GC Column; 30m L, 0.25mm ID, 0.25um thick WZ-01855-04 Mfr # 260W142P
COMPARE
0.25 30 0.25
£881.76GBP / Each
£1,058.11GBP (incl VAT)