This product is now discontinued, View Related Products
Thermo Scientific TRACE TR-1701 (14% Cyanopropylphenyl polysiloxane) GC Columns
Low bleed provides reproducible and accurate results
Thermo Scientific TRACE GC columns have been tested and found to provide less than half the bleed of competitive capillary columns. Low bleeding means fewer problems with low sensitivity, contamination of the detector, lower temperature limits, and shorter life spans. These columns have been manufactured with improved bonding to be extremely robust, minimizing the risk of damage due to contaminated carrier gases. Improved column-to-column reproducibility guarantees that your column will work with the same high standard as those purchased years from now. Each column goes through rigorous testing and are delivered with a certificate of traceability to guarantee the performance.
Thermo Scientific TRACE TR-1701 Columns provide low bleed and high temperature stability, along with alternative selectivity due to its cyano- and phenyl-groups. This alternative selectivity makes these columns an ideal choice for analysis of pesticides.
£821.60 - £946.64GBP / Each
£985.92 - £1,135.97GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (cm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific TRACE 260Q130P GC Column 15-m length, 0.25-mm ID, 0.25µm film thickness WZ-01854-20 Mfr # 260Q130P
COMPARE
0.25
Call For Price
Thermo Scientific TRACE 260Q142P GC Column 30-m length, 0.25-mm ID, 0.25µm film thickness WZ-01854-22 Mfr # 260Q142P
COMPARE
0.25
£821.60GBP / Each
£985.92GBP (incl VAT)
Thermo Scientific TRACE 260Q154P GC Column 60-m length, 0.25-mm ID, 0.25µm film thickness WZ-01854-24 Mfr # 260Q154P
COMPARE
0.25
Call For Price
Thermo Scientific TRACE 260Q131P GC Column 15-m length, 0.32-mm ID, 0.25µm film thickness WZ-01854-26 Mfr # 260Q131P
COMPARE
0.32
Call For Price
Thermo Scientific TRACE 260Q143P GC Column 30-m length, 0.32-mm ID, 0.25µm film thickness WZ-01854-28 Mfr # 260Q143P
COMPARE
0.32
£875.39GBP / Each
£1,050.47GBP (incl VAT)
Thermo Scientific TRACE 260Q155P GC Column; 60m L, 0.32mm ID, 0.25um thick WZ-01854-30 Mfr # 260Q155P
COMPARE
0.32
Call For Price
Thermo Scientific TRACE 260Q224P GC Column 30-m length, 0.32-mm ID, 0.50µm film thickness WZ-01854-32 Mfr # 260Q224P
COMPARE
0.32
Call For Price
Thermo Scientific TRACE 260Q298P GC Column; 30m L, 0.53mm ID, 1.0um thick WZ-01854-34 Mfr # 260Q298P
COMPARE
0.53
£946.64GBP / Each
£1,135.97GBP (incl VAT)
Showing 1- 8 of 8