This product is now discontinued, View Related Products
Thermo Scientific TRACE TR-1 (100% Dimethyl polysiloxane) GC Columns
Low bleed provides reproducible and accurate results
Thermo Scientific TRACE GC columns have been tested and found to provide less than half the bleed of competitive capillary columns. Low bleeding means fewer problems with low sensitivity, contamination of the detector, lower temperature limits, and shorter life spans. These columns have been manufactured with improved bonding to be extremely robust, minimizing the risk of damage due to contaminated carrier gases. Improved column-to-column reproducibility guarantees that your column will work with the same high standard as those purchased years from now. Each column goes through rigorous testing and are delivered with a certificate of traceability to guarantee the performance.
TRACE TR-1 Columns provide general purpose separation on a wide variety of compounds and applications including hydrocarbons, PONA, esters, pesticides, drugs, and amines. With a maximum operating temperature of 320°C and its low phase bleed, these columns are easily baked out and reconditioned. This non-polar column is 100% Dimethyl polysiloxane.
£492.96 - £1,427.96GBP / Each
£591.55 - £1,713.55GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (m) Film Thickness (µm)
Availability Pricing
Thermo Scientific TRACE 260A020P GC Column; 10 m L x 0.10 mm ID x 0.10 um thick WZ-01852-00 Mfr # 260A020P
COMPARE
0.1 10 0.1
Call For Price
Thermo Scientific TRACE 260A227P GC Column; 50 m Lx 0.15 mm ID x 0.50 um thick WZ-01852-02 Mfr # 260A227P
COMPARE
0.15 50 0.5
Call For Price
Thermo Scientific TRACE 260A130P GC Column; 15 m L x 0.25 mm ID x 0.25 um thick WZ-01852-04 Mfr # 260A130P
COMPARE
0.25 15 0.25
£492.96GBP / Each
£591.55GBP (incl VAT)
Thermo Scientific TRACE 260A142P GC Column; 30 m L x 0.25 mm ID x 0.25 um thick WZ-01852-06 Mfr # 260A142P
COMPARE
0.25 30 0.25
£821.60GBP / Each
£985.92GBP (incl VAT)
Thermo Scientific TRACE 260A154P GC Column; 60 m L x 0.25 mm ID x 0.25 um WZ-01852-08 Mfr # 260A154P
COMPARE
0.25 60 0.25
£1,427.96GBP / Each
£1,713.55GBP (incl VAT)
Thermo Scientific TRACE 260A223P GC Column; 30 m L x 0.25 mm ID x 0.50 um WZ-01852-10 Mfr # 260A223P
COMPARE
0.25 30 0.5
£846.31GBP / Each
£1,015.57GBP (incl VAT)
Thermo Scientific TRACE 260A296P GC Column; 30 m L x 0.25 mm ID x 1.0 um thick WZ-01852-12 Mfr # 260A296P
COMPARE
0.25 30 1
£846.31GBP / Each
£1,015.57GBP (incl VAT)
Thermo Scientific TRACE 260A308P GC Column; 60 m L x 0.25 mm ID x 1.0 um thick WZ-01852-14 Mfr # 260A308P
COMPARE
0.25 60 1
£1,427.96GBP / Each
£1,713.55GBP (incl VAT)
Thermo Scientific TRACE 260A131P GC Column; 15 m L x 0.32 mm ID x 0.25 um thick WZ-01852-16 Mfr # 260A131P
COMPARE
0.32 15 0.25
£523.49GBP / Each
£628.19GBP (incl VAT)
Thermo Scientific TRACE 260A143P GC Column; 30 m L x 0.32 mm ID x 0.25 um thick WZ-01852-18 Mfr # 260A143P
COMPARE
0.32 30 0.25
£875.39GBP / Each
£1,050.47GBP (incl VAT)
Thermo Scientific TRACE 260A155P GC Column; 60 m L x 0.32 mm ID x 0.25 um thick WZ-01852-20 Mfr # 260A155P
COMPARE
0.32 60 0.25
Call For Price
Thermo Scientific TRACE 260A224P GC Column; 30 m L x 0.32 mm ID x 0.50 um thick WZ-01852-22 Mfr # 260A224P
COMPARE
0.32 30 0.5
Call For Price
Thermo Scientific TRACE 260A481P GC Column; 60 m L x 0.32 mm ID x 5.0 um thick WZ-01852-24 Mfr # 260A481P
COMPARE
0.32 60 5
Call For Price
Thermo Scientific TRACE 260A297P Column 30-m Length, 0.32-mm ID, 1.00-µm film thickness WZ-01852-26 Mfr # 260A297P
COMPARE
0.32 30 1
£900.11GBP / Each
£1,080.13GBP (incl VAT)
Thermo Scientific TRACE 260A309P GC Column; 60 m L x 0.32 mm ID x 1.0 um thick WZ-01852-28 Mfr # 260A309P
COMPARE
0.32 60 1
Call For Price
Thermo Scientific TRACE 260A395P GC Column; 30 m L x 0.32 mm ID x 3.0 um thick WZ-01852-30 Mfr # 260A395P
COMPARE
0.32 30 3
£900.11GBP / Each
£1,080.13GBP (incl VAT)
Thermo Scientific TRACE 260A213P Column 15-m Length, 0.53-mm ID, 0.50-µm film thickness WZ-01852-32 Mfr # 260A213P
COMPARE
0.53 15 0.5
Call For Price
Thermo Scientific TRACE 260A225P GC Column; 30 m L x 0.53 mm ID x 0.50 um thick WZ-01852-34 Mfr # 260A225P
COMPARE
0.53 30 0.5
Call For Price
Thermo Scientific TRACE 260A396P Column 30-m Length, 0.53-mm ID, 3.00-µm film thickness WZ-01852-36 Mfr # 260A396P
COMPARE
0.53 30 3
Call For Price
Thermo Scientific TRACE 260A470P GC Column; 30 m L x 0.53 mm ID x 5.0 um thick WZ-01852-40 Mfr # 260A470P
COMPARE
0.53 30 5
Call For Price
Showing 1- 20 of 20