Thermo Scientific Orion Star A200 pH/mV/RmV/RDO/DO/BOD/Temperature Benchtop Meter
Durability and dependability make it the ideal choice for every lab, every sample, everyday
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separtely)
Thermo Scientific Orion Star A216 multi-parameter meters offer pH, conductibity and temperature as well as a pH, conductivity, dissolved oxygen and temperature meter. Meters features an intuitive user interface with plain language prompts. A large, backlit graphic LCD shows measurements with temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Up to five point automatic calibration with USA/NIST and Din buffers, auto-buffer recognition, and recalibration feature to fix calibration errors.
Choose between RDO optivcal probe or a polarographic probe for oxygen measurements. Directly measure in mg/L (ppm) or % saturation with automatic temperature, barometric pressure and salinity corrections.
Available as meter only and as complete meter kit configurations.
£3,937.07GBP / Each
£4,724.48GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A216 Star A216 pH/DO Benchtop Meter Kit WZ-58825-42 Mfr # STARA2165
COMPARE
Star A216 pH/DO Benchtop Meter Kit
£3,937.07GBP / Each
£4,724.48GBP (incl VAT)