Thermo Scientific Orion Star A 329 Multiparameter Meter
Advanced features and dependability makes it the top choice for measuring samples
  • Multilanguage interface supports Engliah, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 5000 data points with time date stamp
  • Individual method passwords provide better security
  • Four AA batteries provide over 800 hours of field use, or order optional AC adapter for lab use
Thermo Scientific Orion Star A 300 meters features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
GLP-related features include 5000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
Available as meter only and as complete meter kit configurations
£3,676.74GBP / Each
£4,412.09GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific STAR A329 Star A329pH/ISE/Cond/DO Portable Meter Kit WZ-58825-78 Mfr # STARA3295
COMPARE
Star A329pH/ISE/Cond/DO Portable Meter Kit
£3,676.74GBP / Each
£4,412.09GBP (incl VAT)