Thermo Scientific Orion™ Micro pH Electrodes
Designed to measure samples as small as 0.5 uL in containers as small as 384 well plates
  • Ideal for use with smaller sample sizes and are typically combination electrodes
  • Use micro pH electrodes in conjunction with many instruments including capillary tubes and microwell plates
Microtip fits into small vessels and requires low volumes to obtain pH measurements.
£818.31 - £821.35GBP / Each
£981.97 - £985.62GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 9810BN pH microelectrode, single-junction, 1.3 x 37 mm tip, 3.3-ft WZ-05719-22 Mfr # 9810BN
COMPARE
pH microelectrode, single-junction, 1.3 x 37 mm tip, 3.3-ft
£818.31GBP / Each
£981.97GBP (incl VAT)
Thermo Scientific 9863BN pH microelectrode, Needle Tip, 3.3-ft (1-m) WZ-05719-24 Mfr # 9863BN
COMPARE
pH microelectrode, Needle Tip, 3.3-ft (1-m)
£821.35GBP / Each
£985.62GBP (incl VAT)